
1,1-Dimethylbiguanide hydrochloride, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:1115-70-4
:
Formula:C₄H₁₂ClN₅
MDL Number:MFCD00012582
:165.625 g/mol
Melting Pt:214…224 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- Melting Point:214 - 224 °C