Natural Corn Granules, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Corn is used as pet nutrition and natural pet food.
It contains carbohydrate (77%), protein (8%), fat (2%), fiber (2%) and water (11%).
Caution: Store at ambient temperature.