Order Entry
Germany
ContactUsLinkComponent
Polyvinyl alcohol, M.W. 15000 av, off-white powder
Polyvinyl alcohol, M.W. 15000 av, off-white powder
Supplier:  MP Biomedicals
Certificates
Polyvinyl alcohol, M.W. 15000 av, off-white powder
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • Viscosity (4% aq soln):
    3.5 - 4.5 cps
  • Hydrolysis :
    87.0 - 89.0%
  • Volatiles:
    ≤5%
  • pH:
    4.0 - 6.0(4% aq soln)
  • Ash:
    ≤0.7%

Specification Test Results