Orange G, crystalline powder C.I. 16230
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:1936-15-8
:
Formula:C₁₆H₁₀N₂Na₂O₇S₂
MDL Number:MFCD00012457
:452.38 g/mol
Specification Test Results
- Identity Test :Passes
- UV/VIS: λmax (water):475 - 481 nm