Order Entry
Germany
ContactUsLinkComponent
n-Dodecyl-β-D-maltoside ~97%, white powder
n-Dodecyl-β-D-maltoside ~97%, white powder
Supplier:  MP Biomedicals
Certificates
n-Dodecyl-β-D-maltoside ~97%, white powder
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ~97%
  • FTIR:
    Conforms to Reference
  • Optical Rotation:
    (c=1, methanol) +47 ° ± 5 °

Specification Test Results

Frequently Bought Together