Sodium formate ≥99.0%, white crystalline powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:141-53-7
:
Formula:HCO₂Na
MDL Number:MFCD00013101
:68.01 g/mol
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:≥99.0%
- Insoluble Matter:<0,005%
- Chloride (Cl):<0,001%
- Sulphate (SO4):<0,001%,
- Calcium (Ca):<0,005%
- Heavy Metals (as Pb):<5 ppm
- Iron (Fe):<5 ppm