Sodium bisulphite, powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7631-90-5
:
Formula:HNaO₃S
MDL Number:MFCD00167602
ADR:8,III
:104.06 g/mol
UN:2693
Specification Test Results
- Synonyms:Sodium hydrogensulphite
- Form:White Powder
- Storage:Store at Room Temperature (15 to 30 °C)
- Assay:≥58,5% SO2
- Heavy Metals:≤0,001%
- Insoluble Matter:≤0,005%
- Chloride:≤0,02%
- Iron:≤0,002%