Order Entry
Germany
ContactUsLinkComponent
Potassium sulphate, anhydrous ≥99.0%, powder ACS
Potassium sulphate, anhydrous ≥99.0%, powder ACS
Supplier:  MP Biomedicals
Certificates
Potassium sulphate, anhydrous ≥99.0%, powder ACS
Supplier:  MP Biomedicals

Specification Test Results

  • Synonyms:
    Sulphuric Acid Dipotassium Salt; Sal polychrestum; Arcanum duplicatum; Tartarus vitriolatus
  • Form:
    Powder
  • Storage:
    Store at Room Temperature (15 to 30 °C)
  • Assay:
    ≥99,0% K2SO4
  • Heavy Metals:
    Lead (as Pb): ≤5 ppm; Iron (Fe): ≤5 ppm
  • Insoluble Matter:
    ≤0,01%
  • pH:
    5,5 - 8,5 (5% soln, 25 °C)
  • Chloride (Cl):
    ≤0,001%
  • Nitrogen Compounds (as N):
    ≤5 ppm
  • Calcium (Ca):
    ≤0,01%
  • Magnesium (Mg):
    ≤0,005%
  • Sodium (Na):
    ≤0,02%

Specification Test Results

Frequently Bought Together