Carboxymethyl cellulose sodium salt, white powder, high viscosity
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:9004-32-4
:
Formula:[C₆H₇O₂(OH)x(OCH₂COONa)y]n
MDL Number:MFCD00081472
Melting Pt:> 300 °C (lit.)
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Viscosity (1% aq soln):1000 - 3000 cps