D-(+)-Glucose, anhydrous, white powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:50-99-7
:492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912
Melting Pt:150…152 °C (lit.)
:180.16 g/mol
Specification Test Results
- Identity Test:Passes
- Specific Rotation:52.5 to 53.0°
- Insoluble Matter:<0,005%
- Loss on Drying:<0,20%
- Residue on Ignition:<0,.02%
- Titratable Acid:<0,002 mEq/g
- Chloride:<0,01%
- Sulphate (SO₄) and Sulphite :<0,005%
- Starch:Passes
- Heavy Metals (PB):≤5 ppm,
- Iron:≤5 ppm,