D-(+)-Xylose, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:58-86-6
:
Formula:C₅H₁₀O₅
MDL Number:MFCD00151475
:150.13 g/mol
Melting Pt:155…157 °C (lit.)
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- Optical Rotation:+18° ± 5° (c=4, H2O, NH4OH)
- Loss on Drying:≤0.5%