Polygalacturonic acid (from pectin) ≥86%, powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:25990-10-7
:
MDL Number:MFCD00131972
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:≥86%
- FTIR:Conforms to Reference
- Moisture Content:<10%
- pH (1% susp.):2.5 - 3.5
- Ash Content:≤2%