Order Entry
Germany
ContactUsLinkComponent
4-Aminobenzoic acid, white powder
4-Aminobenzoic acid, white powder
Catalog # ICNA0210256980
Supplier:  MP Biomedicals
CAS Number:  
4-Aminobenzoic acid, white powder
Catalog # ICNA0210256980
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :137.14 g/mol
Density:1.374 g/mL (25 °C) (lit.)
CAS Number:150-13-0
Formula:C₇H₇NO₂
MDL Number:MFCD00007894