L-Arginine hydrochloride, white crystalline powder
:1119-34-2
:
:C₆H₁₅ClN₄O₂
:MFCD00064550
:210.66 g/mol
:Ambient
- Identity Test:Passes
- FTIR:Conforms to Standard
- Optical Rotation (c=8, 2 N HCl):+21.0° to +23°
- Loss on Drying:≤0.5%
- Melting Point:220 - 230 °C