Cesium chloride 99.9%, powder, Optical Grade
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7647-17-8
:
Formula:CsCl
MDL Number:MFCD00010955
:168.36 g/mol
Melting Pt:645 °C (closed cup)
Specification Test Results
- Purity:99,9%
- pH (1% aq soln):5,5 - 6,5
- Barium:≤10 ppm
- Magnesium:≤10 ppm
- Potassium:≤30 ppm
- Iron:≤10 ppm
- Calcium:≤10 ppm
- Solubility (50% aq soln):Clear and Colorless