L(+)-Histidine monohydrochloride monohydrate, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:5934-29-2
:
Formula:C₆H₉N₃O₂·HCl·H₂O
MDL Number:MFCD00151027
:209.63 g/mol
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- pH (2% aq soln):3 - 5
- Loss on Drying (145 - 150°C, 3 hour):7 - 10%
- Optical Rotation:+9,5° ± 1° (c = 1, 6 N HCl)
- pka:1,80 (COOH); 9,33 (NH2); 6,04 (imidazole group) (Lit.)
- Isoelectric Point:7,641(Lit.)