Order Entry
Germany
ContactUsLinkComponent
 
Dextran, M.W. 200000 - 300000, white powder clinical grade
 
Dextran, M.W. 200000 - 300000, white powder clinical grade

 

  • Identity Test:
    Passes
  • pH (1% aq soln):
    4.0-7.0
  • Moisture:
    ≤10%
  • Color by Colorimetric:
    <0.15 abs @ 375 nm
  • Intrinsic Viscosity:
    0.4 - 0.5 (37°C)
  • Sulphated Ash:
    ≤2.0%
  • Cold Water Solubility:
    Passes