Order Entry
Germany
Orders LinkContactUsLinkComponent
Heparin sodium salt, solid

Heparin sodium salt, solid

5 g
Catalog # A3004.0005
Supplier:   PanReac AppliChem
Heparin sodium salt, solid
Catalog # A3004.0005
Supplier:  PanReac AppliChem
CAS Number:  9041-08-1
Heparin sodium salt, solid

5 g

CAS Number:  9041-08-1
Price
438,97 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Reviparin sodium
  • CAS Number:9041-08-1
  • MDL Number:MFCD00081689

Documents

Specifications

Specification Test Results

  • State of Matter:
    Solid
  • Origin:
    Porcine mucosa
  • Storage:
    Store at Room Temperature
  • Assay:
    Min. 150 I.U./mg
  • Solubility (1%; H2O):
    Clear, Colorless
  • pH (1%; H2O; 20 °C):
    5,5 - 8,0
  • Heavy metals (as Pb):
    Max. 0,003%
  • Loss on drying:
    Max. 8%
  • Sodium (Na):
    9,5 - 12,5%

Specifications

  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.