Order Entry
Germany
ContactUsLinkComponent
Guanosine 5'-triphosphate disodium salt ≥98%, white powder
Guanosine 5'-triphosphate disodium salt ≥98%, white powder
Supplier:  MP Biomedicals
Certificates
Guanosine 5'-triphosphate disodium salt ≥98%, white powder
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥98%
  • FTIR:
    Conforms to Standard
  • UV/VIS: λmax (water):
    252 ± 3 nm
  • OD250/OD260:
    1.20 ± 0.08
  • OD280/OD260:
    0.65 ± 0.08

Specification Test Results