Spermidine trihydrochloride ≥99%, crystalline
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:334-50-9
:
Formula:C₇H₂₂Cl₃N₃
MDL Number:MFCD00012918
:254.63 g/mol
Storage Temperature:Freezer
Specification Test Results
- Identity Test:Passes
- Purity:≥99%
- FTIR:Conforms to Standard
- Melting Point:253 - 263 °C