Order Entry
Germany
ContactUsLinkComponent
Potassium dihydrogen phosphate, anhydrous ≥99.0%, crystalline powder
Potassium dihydrogen phosphate, anhydrous ≥99.0%, crystalline powder
Supplier:  MP Biomedicals
Certificates
Potassium dihydrogen phosphate, anhydrous ≥99.0%, crystalline powder
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • Assay:
    ≥99.0%
  • pH (5% aq soln):
    4.1 - 4.5

Specification Test Results