Order Entry
Germany
ContactUsLinkComponent
Coomassie brilliant blue G-250, dark reddish purple powder for electrophoresis C.I. 42655
Coomassie brilliant blue G-250, dark reddish purple powder for electrophoresis C.I. 42655
Catalog # ICNA04808274
Supplier:  MP Biomedicals
CAS Number:  
Coomassie brilliant blue G-250, dark reddish purple powder for electrophoresis C.I. 42655
Catalog # ICNA04808274
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :854.04 g/mol
Storage Temperature:Ambient
CAS Number:6104-58-1
Formula:C₄₇H₄₈N₃NaO₇S₂
MDL Number:MFCD00078482