Orders LinkContactUsLinkComponent
pH indicator, bromophenol blue tablets
pH indicator, bromophenol blue tablets
Supplier:   Lovibond
pH indicator, bromophenol blue tablets
Supplier:  Lovibond
Certificates
pH indicator, bromophenol blue tablets
pH indicator, bromophenol blue tablets
2 Options Available Below
Product Details & Documents
Indicator for pH range 2,8 to 4,4.
Can be used for measuring pH in conjunction with the Comparator 2000+ system and pH disc 2/1B
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 162623A
    Bromophenol blue tablets, pH indicator
    Bromophenol blue tablets, pH indicator
    250 Tab.
    Log in for availability
    Log in for availability
    1 * 250 Tablet
    1 * 250 Tablet - 81,00 €
    81,00 €
    1 * 250 Tablet
       81,00 €
  • LOVI511620
    Bromophenol blue tablets, pH indicator
    Bromophenol blue tablets, pH indicator
    100
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 37,00 €
    37,00 €
    1 * 100 ST
       37,00 €