Cutting machines, hobby cut ³/₄
Supplier: KAISER FOTOTECHNIK
Certificates
About this item
Safety cutting machines with a ground, rotating circular knife, self-sharpening, in a closed plastic head.
- Automatic pressing of the material to be cut at the cutting point
- Transparent pressure ruler
- Two angle systems with cm and inch scales
- Sturdy cutting table made of metal, with printed scales, format markings, degrees and fine grid