Order Entry
Germany
ContactUsLinkComponent
 
Transport Box
  MULC14713
 :  MULTIROIR
Transport Box
  MULC14713
 :  MULTIROIR

 

  • W×D×H ext.:
    482×375×132 mm
  • Description:
    Transport case, M48-132 (cover depth + bottom 60 + 60 mm)
  • Pk:
    4
  • W×D×H int.:
    460×325×120 mm

 

 

This polypropylene case is a solution for securely storing and transporting sensitive equipment, thanks to the option of installing different types of foam.

  • Made of thick polypropylene

  • Excellent stability and high impact resistance
  • Hinges with steel rod

    Supports heavy loads

  • Supplied with convoluted foam, flat foam cutout, or thermoformed tray