ContactUsLinkComponent
Human IgG1 expression control
Human IgG1 expression control
Catalog # MIRUMIR6250
Supplier:  MIRUS
CAS Number:  
undefined
Human IgG1 expression control
Catalog # MIRUMIR6250
Supplier:  MIRUS
CAS Number:  
Specifications
  • Pk:
    100 µG
  • Description:
    Human IgG1 expression control
Specifications
About this item
Human IgG1 Expression Control is a heavy and light chain mixture ideal for verifying antibody expression in mammalian cells.
  • Equimolar ratios of human IgG1 heavy and light chain constructs designed using DNA2.0 IP-Free vector technology
  • Verify human IgG1 expression as a control using ELISA or other methods
  • Suitable for use with any large scale antibody expression system
Delivery: Supplied ready-to-use at 1,0 mg/ml concentration.