Ethanol, anhydrous ≥99%
Catalog # ALCH0310F5
Supplier: Alcoholes Montplet
CAS Number:
Specifications
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Alcoholic grade:> 99.5
- Appearance:Colorless
- Odor:Neutral
Specification Test Results
About this item
:
:46.07 g/mol
Density:>0.780
Storage Temperature:Ambient
UN:1170
ADR:3,III
:GHS02
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568