DNA delivery solution and starter kit, TransIT®-EE
Catalog # MIRUMIR5340
Supplier: MIRUS
CAS Number:
Specifications
- Pk:1
- DNA:Yes
- in vivo:Yes
- Description:TransIT®-EE delivery solution, 40 injections
- Seq:02
Specifications
About this item
TransIT®-EE offers an enhanced expression in vivo delivery solution that delivers DNA to the livers of laboratory mice using hydrodynamic tail vein injection.
- Achieves increased levels of gene expression than compared to other delivery systems and solutions, polylysine and PEI methods
- Suitable for DNA delivery using hydrodynamic tail vein injection to the mouse strain of your choice
Delivery: Each starter kit contains the TransIT®-EE delivery solution, 10 syringes, needles and alcohol swabs, and a conical plastic tube to restrain the mouse while tail vein injection is performed. The TransIT®-EE delivery solution is also available separately.