Expression system, CHOgro®
Catalog # MIRUMIR-6260
Supplier: MIRUS
CAS Number:
Specifications
- Pk:1 KIT
- Description:CHOgro® expression system
- Seq:01
Specifications
About this item
CHOgro® offers a high titre transient transfection system for suspension CHO cells.
- Enables high protein titres with simple workflow
- Quick adaptation to CHO cell lineages
- High density growth with minimal cell clumping post transfection
- No commercial license required; animal-origin-free
Delivery: Kit contains CHOgro® expression medium, 2 L; TransIT-PRO® transfection reagent, 1 ml; CHOgro® complex formation solution, 100 ml; Poloxamer 188 solution,100 ml and L-glutamine, 100 ml. All components can also be purchased separately.