Order Entry
Germany
Orders LinkContactUsLinkComponent
Ethanol absolute ≥99%

Ethanol absolute ≥99%

Flammable
Irritant
Supplier:   KLINIPATH
Ethanol absolute ≥99%
Supplier:  KLINIPATH
Certificates
Ethanol absolute ≥99%

1 Options Available Below

Product Details & Documents
Synonyms:  Ethyl alcohol
  • CAS Number:64-17-5
  • Formula:C₂H₅OH
  • MDL Number:MFCD00003568
  • Molecular Weight:46.07 g/mol
  • Flash Pt:12 °C (closed cup)
  • Storage Temperature:Ambient
  • UN:1170
  • ADR:3,II

Documents

Certificates
Search by Lot Number
Product Family Options

Product Information