Order Entry
Germany
ContactUsLinkComponent
Ethanol denatured ≥99%, KLINIPATH for histology (denatured with 5% methanol)
Ethanol denatured ≥99%, KLINIPATH for histology (denatured with 5% methanol)
Catalog # VWRK4098-9020
Supplier:  KLINIPATH
CAS Number:  
Ethanol denatured ≥99%, KLINIPATH for histology (denatured with 5% methanol)
Catalog # VWRK4098-9020
Supplier:  KLINIPATH
CAS Number:  

Specifications

  • Pk:
    20 L
  • Seq:
    5302
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (Ethanol) (V/V):
    94 - 97.0 %
  • Methanol (V/V):
    3.0 - 5.5%
  • Water (V/V):
    Max. 0.3%

Specification Test Results

About this item

Synonyms: Ethyl alcohol denatured
 :
 :46.07 g/mol
Storage Temperature:Ambient
UN:1170
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
CAS Number:64-17-5
Formula:H₃CCH₂OH
MDL Number:MFCD00003568