Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Agarose

100 g
Catalog # ICNA04800259
Supplier:   MP Biomedicals
Agarose
Catalog # ICNA04800259
Supplier:  MP Biomedicals
CAS Number:  9012-36-6
Agarose
Agarose

100 g

CAS Number:  9012-36-6
Price
652,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Low gel temperature electroendosmosis: 0,1 gel strength 1,5% solution: ≥550 gm/cm2 gel temperature (1%): 30 °C A 1% solution remains fluid at 37 °C for up to 24 hours. Will set to a firm gel at 25 °C, and not remelt until temperatures exceed 65 °C. This ability to remain in solution at 30 to 37 °C allows a second digest on a restriction enzyme fragment without need to recover it from the gel. Agarose LGT is also preferred for studies of cellular antibody production and the Jerne Plaque assay of immune responses. Also used in agarose droplet macrophage inhibitory factor (MIF) assay.

  • CAS Number:9012-36-6
  • MDL Number:MFCD00081294
Specifications

Specifications

  • Pk:
    100 g

Specifications