Anti-CD22 Mouse Monoclonal Antibody (CF-Blue) [clone: HIB22]
Catalog # ABNOMAB13849
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:CD22
- Clonality:Monoclonal
- Clone:HIB22
- Conjugation:CF-Blue™
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Gene ID:933
- Size:100 reactions
- Pk:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against human CD22.
Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: HIB22
Conjugation: CF-Blue
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human