Anti-ITGAL Mouse Monoclonal Antibody (CF-Blue) [clone: HI111]
Catalog # ABNOMAB13795
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:ITGAL
- Clonality:Monoclonal
- Clone:HI111
- Conjugation:CF-Blue™
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Gene ID:3683
- Size:100 reactions
- Pk:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against native human ITGAL.
Type: Primary
Antigen: ITGAL
Clonality: Monoclonal
Clone: HI111
Conjugation: CF-Blue
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human,Mouse