Order Entry
Germany
ContactUsLinkComponent
Anti-CD45RB Mouse Monoclonal Antibody (CF-Blue) [clone: MC5/2]
Anti-CD45RB Mouse Monoclonal Antibody (CF-Blue) [clone: MC5/2]
  ABNOMAB13683
 :  Abnova
Anti-CD45RB Mouse Monoclonal Antibody (CF-Blue) [clone: MC5/2]
  ABNOMAB13683
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen symbol:
    CD45RB
  • Clonality:
    Monoclonal
  • Clone:
    MC5/2
  • Conjugation:
    CF-Blue™
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Gene ID:
    5788
  • Size:
    100 reactions
  • Pk:
    100 Assays

 

 

Mouse monoclonal antibody raised against human CD45RB.

Type: Primary
Antigen: CD45RB
Clonality: Monoclonal
Clone: MC5/2
Conjugation: CF-Blue
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human