Order Entry
Germany
ContactUsLinkComponent
G418 disulphate 50 mg/ml in water
G418 disulphate 50 mg/ml in water
Catalog # MIRUMIR5920
Supplier:  MIRUS
CAS Number:  
G418 disulphate 50 mg/ml in water
Catalog # MIRUMIR5920
Supplier:  MIRUS
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :692.72 g/mol
CAS Number:108321-42-2
Formula:C₂₀H₄₄N₄O₁₈S₂
MDL Number:MFCD00058314