Plasmid delivery controls, Label IT®
Supplier: MIRUS
Certificates
About this item
Fluorescently labelled plasmids ideal for in vitro and in vivo tracking.
- For in vivo delivery
- Easily visualise transfected cells and assess delivery efficiency using fluorescent microscopy
- Suitable for co-delivery experiments with functional plasmids
Delivery: Supplied as a ready to use 0,5 mg/ml concentrate.