siRNA tracker intracellular localization kits, Label IT®
Supplier: MIRUS
Certificates
About this item
Label IT® siRNA Tracker Intracellular Localization Kits facilitate efficient, direct labeling for in vitro and in vivo tracking of siRNA.
- Label up to 50 μg siRNA
- Easily and consistently control the labeling reactions and labeling density
- High efficiency labeling with optimal visualisation of siRNA in cells and tissues
Delivery: Each kit contains Label IT® siRNA Tracker Reagent, Label IT® Reconstitution Solution, 10X Labeling Buffer A, and siRNA Dilution Buffer.