ContactUsLinkComponent
High visibility sleeve protectors, cut resistant, HyFlex® 11-202 HI-VIZ™
High visibility sleeve protectors, cut resistant, HyFlex® 11-202 HI-VIZ™
Catalog # ANSE11-202
Supplier:  Ansell
undefined
High visibility sleeve protectors, cut resistant, HyFlex® 11-202 HI-VIZ™
Catalog # ANSE11-202
Supplier:  Ansell
Specifications
  • Garment fit:
    Regular
  • Type:
    High visibility sleeve protectors
  • Garment application:
    Mechanical protection
  • Gender:
    Unisex
  • Risk area:
    Cuts,
    HiVis
  • Colour:
    Hi-vis yellow
  • Length:
    475 mm
  • Fabric:
    polyester, nylon and glass
  • Pk:
    12
Specifications
About this item
This sleeve made of polyester, nylon and glass fibre has a level EN CUT 5 performance and an increased abrasion protection. Safety is also enhanced through its visibility colour and level 1 thermal protection.
  • Sleeve delivers the best comfort through its anatomical design
  • Offers an improved fit thanks to its adjustable VELCRO® strap system
  • Freedom of movement leads to increased productivity and worker acceptance
Ideal for use in body shops (excluding welding), for glass and metal sheet handling and for stamping.
Gauge: 13
Certifications: EN 388 (2542), EN 407 (X1XXXX); Cat. III