Order Entry
Germany
ContactUsLinkComponent
Anti-CD45RO Mouse Monoclonal Antibody (APC-Cyanine7) [clone: UCHL1]
Anti-CD45RO Mouse Monoclonal Antibody (APC-Cyanine7) [clone: UCHL1]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD45RO Mouse Monoclonal Antibody (APC-Cyanine7) [clone: UCHL1]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 25-0457-T025
  • 25-0457-T100
  • Antigen symbol:
  • CD45RO
  • CD45RO
  • Antigen name:
  • CD45 isoform known as CD45RO, a protein tyrosine phosphatase
  • CD45 isoform known as CD45RO, a protein tyrosine phosphatase
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)-Cy7®
  • APC (Allophycocyanin)-Cy7®
  • Clone:
  • UCHL1
  • UCHL1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 5788
  • 5788
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • +2 °C to +8 °C, protected from light
  • +2 °C to +8 °C, protected from light

Specifications

Related Information