Anti-CD20 Mouse Monoclonal Antibody (APC-Cyanine7) [clone: 2H7]
The 2H7 antibody reacts with human CD20, expressed by pre-B cells, resting and activated B cells, follicular dendritic cells and a small subset of T cells, but not on plasma cells. CD20 is a 33-37 kDa transmembrane protein which is differentially phosphorylated in resting and active B cells. The CD20 molecule plays a role in regulating B-cell activation. The 2H7 antibody is reported to be cross-reactive with several non-human species including Baboon, Chimpanzee, Cynomolgus and Rhesus. Please choose the appropriate format for each application.
Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 2H7
Conjugation: APC-Cyanine7
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
- Catalog No:
- 25-0209-T025
- 25-0209-T100
- Antigen symbol:
- CD20
- CD20
- Antigen synonyms:
- Bp35|B1|Leu-16
- Bp35|B1|Leu-16
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)-Cy7®
- APC (Allophycocyanin)-Cy7®
- Clone:
- 2H7
- 2H7
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 931
- 931
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- +2 °C to +8 °C, protected from light
- +2 °C to +8 °C, protected from light