Armenian Hamster IgG Isotype Control [clone: PIP] (APC (Allophycocyanin))
20-4888-U100
- Antibody type:Control
- Clonality:Monoclonal
- Clone:PIP
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Armenian hamster
- Isotype:IgG
- Storage temperature:+2 °C to +8 °C, protected from light
- Size:100 μg
- Pk:100 µG
The PIP immunoglobulin is useful as an isotype-matched control. The PIP immunolglobulin is specific for Glutathione S-transferase and can be used as an isotype control for Armenian Hamster IgG antibodies.
Type: Control
Antigen: Isotype Control
Clonality: Monoclonal
Clone: PIP
Conjugation: APC
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: N/A