Order Entry
Germany
ContactUsLinkComponent
Anti-SYP Rabbit Polyclonal Antibody
Anti-SYP Rabbit Polyclonal Antibody
Supplier:  IMMUNOLOGIC
Certificates
Anti-SYP Rabbit Polyclonal Antibody
Supplier:  IMMUNOLOGIC

Specifications

  • Catalog No:
  • ILP11120-C01
  • ILP11120-C05
  • VWRKILP11120-C1
  • Antigen symbol:
  • SYP
  • SYP
  • SYP
  • Antigen name:
  • Synaptophysin
  • Synaptophysin
  • Synaptophysin
  • Antigen synonyms:
  • MRX96
  • MRX96
  • MRX96
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Gene ID:
  • 6855
  • 6855
  • 6855
  • Isotype:
  • IgG
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Form:
  • Concentrated
  • Concentrated
  • Concentrated
  • Storage temperature:
  • 2°C - 8°C
  • 2°C - 8°C
  • 2°C - 8°C
  • Tested applications:
  • Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections
  • Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections
  • Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections

Specifications

Related Information

Frequently Bought Together