About this item
Ki-67 protein is a nuclear protein doublet plays a pivotal role in maintaining cell proliferation. In diagnostic histopathology and cell biology, the antibody has proven valuable for the demonstration of the Ki-67 antigen in normal and neoplastic cells, for example in soft-tissue sarcoma, prostatic adenocarcinoma, and breast carcinoma. It has been confirmed as a very powerful single prognostic factor for overall survival, with highly proliferative cases showing a much poorer outcome than tumors with low proliferation. In breast cancer, the proliferative index measured by Ki-67 immunoreactivity has both prognostic and predictive value.
- Control: Tonsil
- Dilution Range: 1:400-1:800 for immunohistochemical staining
Pretreatment: Heat induced epitope retrieval in 10 mM citrate buffer , pH 6,0 for 20 minutes is required for IHC staining on formalin-fixed, paraffin embedded tissue sections.
Type: Primary
Antigen: MKI67
Clonality: Monoclonal
Clone: MIB-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- ILM9252-C01
- ILM9252-C05
- ILM9252-C1
- Antigen symbol:
- MKI67
- MKI67
- MKI67
- Antigen name:
- Marker of proliferation Ki-67
- Marker of proliferation Ki-67
- Marker of proliferation Ki-67
- Antigen synonyms:
- MIB-|PPP1R105
- MIB-|PPP1R105
- MIB-|PPP1R105
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- MIB-1
- MIB-1
- MIB-1
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 4288
- 4288
- 4288
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Form:
- Concentrated
- Concentrated
- Concentrated
- Molecular weight:
- 345 - 395 kDa
- 345 - 395 kDa
- 345 - 395 kDa
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
- Tested applications:
- Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections
- Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections
- Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections