Washdown resistant labels, ToughWash™
Supplier: Brady
Certificates
About this item
The Toughwash™ washdown resistant labels are a durable, print on demand pressure sensitve adhesive product.
- Protection from harsh washdown environments
- Tested at washdown temperatures up to 80 °C and pressures up to 1500 psi
Ideal for the food and beverage processing industry.