Inspection mirror
Stainless steel, electrolytically polished inspection mirror, 152×152 mm
MICRQDIST-66P
Inspection Mirror has a swivel connection allowing it to be attached to stainless steel handle and allows the operator to run the unit behind equipment and fixtures to ensure all surfaces have been properly cleaned.
- Highly polished stainless steel
- A 360° swivel allows optimum flexibility
- Ideal for hard to reach areas
Ordering information: The optional flashlight attachment is ideal for use in tanks and other low light applications. Rod is not included, but fits to any SSU series handle.
- Description:Stainless steel, electrolytically polished inspection mirror
- Length:152×152 mm
Accessories