Order Entry
Germany
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS) 0.01 N in aqueous solution
Sodium dodecyl sulphate (SDS) 0.01 N in aqueous solution
Catalog # KRAF14141.1000
Supplier:  Bernd Kraft
CAS Number:  
undefined
Sodium dodecyl sulphate (SDS) 0.01 N in aqueous solution
Catalog # KRAF14141.1000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
 :288.38 g/mol
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175