PIG® clear cube bag spill kit
Supplier: NEW PIG
Certificates
About this item
This "Grab & go" spill response kit has a see-through design for easy inventory of spill response supplies.
- Durable see-through cube bag that makes it easy to check spill response supplies with a quick
- Hook and loop box-lid closure protects the contents and is easy to open for access to absorbents
- Shoulder strap affords comfortable transport to the spill site and makes kit ideal for conveniently hanging on a wall
- PIG HazMat absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
- See-through bag with bold Spill Kit lettering for quick identification
Contains temporary disposal bags and ties for neat and convenient disposal of used absorbents.
Ordering information: Includes PIG® socks to contain and absorb spills, PIG® mats to absorb the contained liquid and PIG® pillows for larger spills and persistent drips; suitable for use on chemical spills.
Caution: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.