ContactUsLinkComponent
Vane wheel for heat/air flow
Vane wheel for heat/air flow
Catalog # BINH1003101
Supplier:  3B Scientific
undefined
Vane wheel for heat/air flow
Catalog # BINH1003101
Supplier:  3B Scientific
Specifications
  • Description:
    Vane wheel
  • Material:
    Aluminium
  • Pk:
    1
Specifications
About this item
Vane wheel device is used for demonstrating air and heat flows generated by a burning candle, water vapour or other heat sources. Made of Aluminium and mountable on a long needle.