Evacuable pellet dies, Atlas™
Supplier: SPECAC
Certificates
About this item
These evacuable pellet dies are designed for compacting powdered samples into discs or briquettes.
- Can be used to produce sample pellets from 5 to 40 mm diameter
- Provides pellets with clarity and quality
- Can cover a range of load capabilities
- An evacuation port is provided to remove the moisture during pressing process
Suited for the preparation of solid KBr pellets for FT-IR molecular spectroscopic analysis and XRF pellets for X-Ray fluorescence atomic spectroscopic analysis.
Delivery: Supplied with evacuable base, body, plunger, a pair of internal pellet pressing surfaces, extractor ring, and O-ring kit.